Dipropylamin 99%
Glasflasche, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 142-84-7 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: 7°(45°F) Refractive Index: 1.4050 Odour: Ammonia-like Melting Point: -63° Boiling Point: 109-110° Beilstein Reference: 505974 Merck Reference: 14,3343 UN No.: 2383 MDL No.: MFCD00009362
Synonyms: N-Propyl-1-propanamine
- CAS Number:142-84-7
- Formula:C₆H₁₅N
- MDL Number:MFCD00009362
- Molecular Weight:101.19 g/mol
- Merck Index:14,3343
- Boiling Pt:109…110 °C
- Melting Pt:–63 °C
- Density:0,738
- Flash Pt:7 °C (45 °F)
- Storage Temperature:Raumtemperatur
- EINECS:205-565-9
- UN:2383
- ADR:3,II
Specifications
Specifications
- Packung:Glasflasche
- VE:100 mL



