Methylacetat ≥99%
Glasflasche, 500 mL
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 79-20-9 Molecular Formula: CH3COOCH3 Formula Weight: 74.08 Flash Point: -10°(14°F) Refractive Index: 1.3620 Odour: Pleasant Melting Point: -98° Boiling Point: 56-58° Beilstein Reference: 1736662 Merck Reference: 14,6008 UN No.: 1231 MDL No.: MFCD00008711
Synonyme: Acetic acid methyl ester
- CAS Nummer:79-20-9
- Formel:C₃H₆O₂
- MDL-Nummer:MFCD00008711
- Molekulargewicht:74.08 g/mol
- Merck-Index:14,6008
- Siedepunkt:56…58 °C
- Schmelzender Platingehalt:–98 °C
- Dichte:0,934
- Blitz Pt:–10 °C (14 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:201-185-2
- UN:1231
- ADR:3,II
Technische Daten
Technische Daten
- Packung:Glasflasche
- VE:500 mL
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



