Domiphen bromide 97%
100 g
Purity: 97% CAS No.: 538-71-6 Molecular Formula: C22H40BrNO Formula Weight: 414.48 Melting Point: 117-118° Beilstein Reference: 3922742 MDL No.: MFCD00011769
: (Dodecyldimethyl-2-phenoxyethyl)ammonium bromide, N,N-Dimethyl-N-(2-phenoxyethyl)dodecan-1-aminium bromide
- :538-71-6
- :C₂₂H₄₀BrNO
- :MFCD00011769
- :414.48 g/mol
- :14,3415
- :117…118 °C
- :Raumtemperatur
- :208-702-0
- VE:100 g