Silberchlorid ≥95%
500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 95% CAS No.: 7783-90-6 Molecular Formula: AgCl Formula Weight: 143.32 Physical Form: Coarse Powder Density: 5.56 Refractive Index: 2.071 Sensitivity: Light Sensitive Appearance: White Odour: Odourless Melting Point: 455° Boiling Point: 1550° Solubility: Practically insoluble in water. Soluble in ammonia, solutions of alkali cyanides, thiosulfates, ammonium carbonates. Insoluble in alcohol, dilute acids Merck Reference: 14,8509 UN No.: 3077 MDL No.: MFCD00003399
Silver plating
- CAS Nummer:7783-90-6
- Formel:AgCl
- MDL-Nummer:MFCD00003399
- Molekulargewicht:143.32 g/mol
- Merck-Index:14,8509
- Siedepunkt:1550 °C
- Schmelzender Platingehalt:455 °C
- Dichte:5,56
- Lagertemperatur:Raumtemperatur
- EINECS:232-033-3
- ADR:9,III
Documents
Technische Daten
Technische Daten
- VE:500 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



