Ninhydrin ACS
Glass bottle for solids, 100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
CAS No.: 485-47-2 Molecular Formula: C9H6O4 Formula Weight: 178.14 Physical Form: Crystalline Sensitivity: Light Sensitive Appearance: Pale yellow to pale brown Melting Point: 251° dec. Solubility: Soluble in water and alcohol Beilstein Reference: 1910963 Merck Reference: 14,6554 MDL No.: MFCD00003791
Reagent for determination of amino acids and amine containing compounds, as an intermediate.
Synonyme: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
- CAS Nummer:485-47-2
- Formel:C₉H₆O₄
- MDL-Nummer:MFCD00003791
- Molekulargewicht:178.14 g/mol
- Merck-Index:14,6554
- Siedepunkt:351 °C (1013 hPa)
- Schmelzender Platingehalt:251 °C (Zersetzung)
- Dichte:0,862 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:207-618-1
Dokumente
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:100 g



