(±)-Epoxystyrol ≥98%
Glass bottle for solids, 100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98+% CAS No.: 96-09-3 Molecular Formula: C8H8O Formula Weight: 120.15 Storage Temperature: 2°C to 8°C Flash Point: 74°(165°F) Refractive Index: 1.5350 Melting Point: -37° Boiling Point: 193-196° Beilstein Reference: 108582 UN No.: 2810 MDL No.: MFCD00005121
Synonyme: Styrene oxide
- CAS Nummer:96-09-3
- Formel:C₈H₈O
- MDL-Nummer:MFCD00005121
- Molekulargewicht:120.15 g/mol
- Siedepunkt:193…196 °C
- Schmelzender Platingehalt:–37 °C
- Dichte:1,053
- Blitz Pt:74 °C (165 °F)
- EINECS:202-476-7
- UN:2810
- ADR:6.1,III
Dokumente
Zertifikate
Suche nach Chargennummermsds_ALFAL07821
Sicherheitsdatenblatt (SDS)
Organic Building Blocks and Intermediates
Anwendungshinweise
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



