Methyllinoleat 95%
Glasflasche für Feststoffe, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 95% CAS No.: 112-63-0 Molecular Formula: C19H34O2 Formula Weight: 294.47 Storage Temperature: 2°C to 8°C Flash Point: 200°(392°F) Refractive Index: 1.4640 Melting Point: -35° Boiling Point: 207-208°/11mm Beilstein Reference: 1727614 Merck Reference: 14,6095 MDL No.: MFCD00009534
Synonyms: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester
- CAS Number:112-63-0
- Formula:C₁₉H₃₄O₂
- MDL Number:MFCD00009534
- Molecular Weight:294.48 g/mol
- Merck Index:14,6095
- Boiling Pt:207…208 °C (11 mmHg)
- Melting Pt:–35 °C
- Density:0,888
- Flash Pt:200 °C (392 °F)
- EINECS:203-993-0
Documents
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



