Accessories for comparators
Chlorine, 3/40C, 1,5-3,5 mg/l
Katalognr. LOVI234030
Anbieter: Lovibond
Direkt zu
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Technische Daten
- Messbereich:1,5-3,5 mg/l
- Seq:17
- Code:3/40C
- Testmethode:Chlorine
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.