Accessories for comparators
Chlorine, 3/40N, 1,1-2,0 mg/l
Katalognr. LOVI233960
Anbieter: Lovibond
Direkt zu
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Technische Daten
- Messbereich:1,1-2,0 mg/l
- Seq:15
- Code:3/40N
- Testmethode:Chlorine
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.