AuftragsverfolgungKontakt
Accessories for comparators | Avantor
Accessories for comparators
Silica, 3/140, 0,1 - 1,0 mg/l
Katalognr. LOVI230250
Anbieter:   Lovibond
Accessories for comparators
Katalognr. LOVI230250
Anbieter:  Lovibond
Accessories for comparators
Accessories for comparators
Silica, 3/140, 0,1 - 1,0 mg/l
Preis
406.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Technische Daten
  • Messbereich:
    0,1 - 1,0 mg/l
  • Seq:
    73
  • Code:
    3/140
  • Testmethode:
    Silica