AuftragsverfolgungKontakt
Stoppers, with drip tip, MBL® | Avantor
Stoppers, with drip tip, MBL®
12/21, With pointed bottom
Katalognr. BIBBTRF692
Anbieter:   SciLabware
Stoppers, with drip tip, MBL®
Katalognr. BIBBTRF692
Anbieter:  SciLabware
Stoppers, with drip tip, MBL®
Stoppers, with drip tip, MBL®
12/21, With pointed bottom
Preis
SFr. 71.50
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
Ideally suited for use with volumetric flasks etc. Lower closed end of stopper features a 'drip-tip' to ensure adhered liquid is returned to the flask. Flat head allows stopper to be stood on end, thus preventing contamination of the ground surface.
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Hollow-blown to reduce weight
Technische Daten
  • NS:
    12/21
  • Typ:
    With pointed bottom
  • Seq:
    03