Büchner-Trichter
95 mm, 500 ml, 2
Katalognr. BIBB3710/04
Anbieter: SciLabware
Direkt zu
Product Details & Documents
PYREX®, borosilicate glass, with sintered glass disc.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
For convenient filtration of solutions.
Technische Daten
- Platte Ø:95 mm
- Porosität:2
- Kapazität:500 ml
- Seq:38



