Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Büchner-Trichter

60 mm, 125 ml, 5
Katalognr. BIBB3700/10
Anbieter:   SciLabware
Büchner-Trichter
Katalognr. BIBB3700/10
Anbieter:  SciLabware
Büchner-Trichter
Büchner-Trichter

60 mm, 125 ml, 5

Preis
SFr. 1’910.00
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents

PYREX®, borosilicate glass, with sintered glass disc.

  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance

For convenient filtration of solutions.

Technische Daten
  • Platte Ø:
    60 mm
  • Porosität:
    5
  • Kapazität:
    125 ml
  • Seq:
    32
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.