Büchner-Trichter
60 mm, 125 ml, 3
Katalognr. BIBB3700/06
Anbieter: SciLabware
Direkt zu
Product Details & Documents
PYREX®, borosilicate glass, with sintered glass disc.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
For convenient filtration of solutions.
Technische Daten
- Platte Ø:60 mm
- Porosität:3
- Kapazität:125 ml
- Seq:28



