Büchner-Trichter
40 mm, 50 ml, 0
Katalognr. BIBB3630/01
Anbieter: SciLabware
Direkt zu
Product Details & Documents
PYREX®, borosilicate glass, with sintered glass disc.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
For convenient filtration of solutions.
Technische Daten
- Platte Ø:40 mm
- Porosität:0
- Kapazität:50 ml
- Seq:10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



