Microplates, 96-well, non binding
96-well microplate, U-bottom, non binding, solid bottom, Klar
Catalog # 738-0165
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
Specifications
- Farbe:Klar
- Bezeichnung:96-well microplate, U-bottom, non binding, solid bottom
- Seq:01
- Cat. No.:738-0165
- Umweltzertifizierung:ISO 14001 (241002); ISO 50001 (241002)
- Umweltfreundlichkeit:Umweltschonende Herstellung