Quarz-Kontrollplatten
Product Details & Documents
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
- Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
- Available in three optical rotation values
- Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
Zertifizierung: Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
Bestellinformationen: Sadlles can be ordered in option.
Lieferinformationen: Supplied with a certificate of verification in a plastic storage case.
Technische Daten
Technische Daten
- VE:1
- Bezeichnung:Thermoblock für Quarz-Kontrollplatten