Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Gentamicinsulfat

Gentamicinsulfat

5 g
Katalognr. P4020-5GR
Anbieter:   Biowest
Gentamicinsulfat
Katalognr. P4020-5GR
Anbieter:  Biowest
CAS Nummer:  1405-41-0
Gentamicinsulfat
Gentamicinsulfat

5 g

CAS Nummer:  1405-41-0
Preis
SFr. 91.80
Einloggen für Verfügbarkeit
1 * 5 g
Product Details & Documents

Gentamycin sulphate acts against Gram-negative, Gram-positive bacteria and mycoplasma.

  • Storage at 2 to 8 °C
  • Shelf life of 48 months

Used in cell culture applications at 1 ml/l. This concentration is for tissue culture media containing serum; serum-free media generally require lower concentration.

Caution: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Synonyme:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Nummer:1405-41-0
  • MDL-Nummer:MFCD00270181

Documents

Technische Daten

Technische Daten

  • VE:
    5 g
  • Cat. No.:
    P4020-5GR

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.