Gentamicinsulfat
1 g
Anbieter: Biowest
Direkt zu
Product Details & Documents
Gentamycin sulphate acts against Gram-negative, Gram-positive bacteria and mycoplasma.
- Storage at 2 to 8 °C
- Shelf life of 48 months
Used in cell culture applications at 1 ml/l. This concentration is for tissue culture media containing serum; serum-free media generally require lower concentration.
Caution: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.
Synonyme: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
- CAS Nummer:1405-41-0
- MDL-Nummer:MFCD00270181
Documents
Zertifikate
Suche nach ChargennummerTechnische Daten
Technische Daten
- VE:1 g



