Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Trypsin

Trypsin

Glass bottle for solids, 500 mg
  EURLEMR308500
 :   EUROCLONE
Trypsin
  EURLEMR308500
 :  EUROCLONE
 :  
Trypsin

Glass bottle for solids, 500 mg

 :  
 
 
   
1 * 500 mg
 
 

 

  • Packung:
    Glass bottle for solids
  • VE:
    500 mg
  • Cat. No.:
    EURLEMR308500
  • CAS Nr.:
    9002-07-7
  • Enzymname:
    Trypsin
  • MDL No.:
    MFCD00082094
  • Lagerbedingungen:
    Refrigerator

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.