Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Trypsin

Trypsin

1 KIT
Catalog # EURLEMR308001
Supplier:   EUROCLONE
Trypsin
Catalog # EURLEMR308001
Supplier:  EUROCLONE
CAS Number:  
Trypsin

1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications

Specifications

  • VE:
    1 KIT
  • Cat. No.:
    EURLEMR308001
  • CAS Nr.:
    9002-07-7
  • Enzymname:
    Trypsin
  • MDL No.:
    MFCD00082094
  • Lagerbedingungen:
    Refrigerator

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.