Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
L-thyroxine, Pulver

L-thyroxine, Pulver

100 mg
Katalognr. ICNA02152145.1
Anbieter:   MP Biomedicals
L-thyroxine, Pulver
Katalognr. ICNA02152145.1
Anbieter:  MP Biomedicals
CAS Nummer:  51-48-9
L-thyroxine, Pulver

100 mg

CAS Nummer:  51-48-9
Preis
SFr. 46.60
Einloggen für Verfügbarkeit
1 * 100 mg
Product Details & Documents

Soluble in solutions of alkali hydroxides (50 mg/ml in 4 M ammonium hydroxide in methanol); slightly insoluble in alcohol; insoluble in water, acetone, chloroform and ether.

Synonyme:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS Nummer:51-48-9
  • Formel:C₁₅H₁₁I₄NO₄
  • MDL-Nummer:MFCD00002595
  • Molekulargewicht:776.874 g/mol
Technische Daten

Technische Daten

  • VE:
    100 mg

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.