Methyloctanoat
Glasflasche für Feststoffe, 1 g
Katalognr. BTRN1199
Direkt zu
Artikel gestrichen
Product Details & Documents
Synonyme: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
- CAS Nummer:111-11-5
- Formel:C₉H₁₈O₂
- MDL-Nummer:MFCD00009551
- Molekulargewicht:158.24 g/mol
- Siedepunkt:190 °C (1013 hPa)
- Schmelzender Platingehalt:–36,7 °C
- Dichte:0,877 g/cm³ (20 °C)
- Blitz Pt:68 °C
- EINECS:203-835-0
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



