Arachidonsäure
Glasflasche für Feststoffe, 100 mg
Catalog # BTRN1042
Jump to:
Discontinued
Product Details & Documents
Synonyms: (5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid
- CAS Number:506-32-1
- Formula:C₂₀H₃₂O₂
- MDL Number:MFCD00004417
- Molecular Weight:304.5 g/mol
- EINECS:208-033-4
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



