Methyldocosanoat
Glasflasche für Feststoffe, 500 mg
BTRN1036
: Methyl behenate, Docosanoic acid methyl ester, Behenic acid methyl ester
- :929-77-1
- :C₂₃H₄₆O₂
- :MFCD00009347
- :354.62 g/mol
- :213-207-8
- Packung:Glasflasche für Feststoffe
- VE:500 mg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.