Methylpalmitat
Glasflasche für Feststoffe, 1 g
Katalognr. BTRN1015
Direkt zu
Artikel gestrichen
Product Details & Documents
Synonyme: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester
- CAS Nummer:112-39-0
- Formel:C₁₇H₃₄O₂
- MDL-Nummer:MFCD00008994
- Molekulargewicht:270.46 g/mol
- Siedepunkt:417 °C (1013 hPa)
- Schmelzender Platingehalt:29,9 °C
- Dichte:0,8804 g/cm³ (20 °C)
- EINECS:203-966-3
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



