L(+)-Lysin Monohydrochlorid
Glasflasche für Feststoffe, 500 g
Katalognr. ICNA0219469790
Anbieter: MP Biomedicals
Direkt zu
Product Details & Documents
Storage: Store at room temperature (15-30 °C)
L-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine. L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.
L-Lysine monohydrochloride is a key amino acid in calcium absorption.
- Cell Culture Reagent Grade
- Presentation: White Powder
- Soluble in aqueous solution (10 mg/mL-clear, colorless solution).
Synonyme: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl
- CAS Nummer:657-27-2
- Formel:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
- MDL-Nummer:MFCD00064564
- Molekulargewicht:182.65 g/mol
- Merck-Index:13,05656
- Siedepunkt:290 °C (1013 hPa)
- Schmelzender Platingehalt:263 °C (Lit.)
- Lagertemperatur:Raumtemperatur
- EINECS:200-294-2
Technische Daten
Ergebnisse des Spezifikationstests
- Identity Test:Passes
- Purity:≥99%
- pH (10% aq soln):5-6
- Loss on Drying:≤1.0%
- Optical Rotation:+20° to +23° (c=8, 2N HCl)
- Heavy Metals:≤15ppm
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:500 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



