Prepared microscope slides: typical monocot roots
Convallaria (ts), lily of valley showing monocot root of endodermis, pericycle, phloem, xylem
Catalog # LIEDAS217C
Supplier: LIEDER JOHANNES
Jump to:
Product Details & Documents
Prepared microscope slides in systematic order showing the anatomy of typical monocot roots in angiospermae.
(ts) total structure
Specifications
- Bezeichnung:Convallaria (ts), lily of valley showing monocot root of endodermis, pericycle, phloem, xylem