Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Prepared microscope slides: typical monocot roots
Prepared microscope slides: typical monocot roots
Convallaria (ts), lily of valley showing monocot root of endodermis, pericycle, phloem, xylem
Catalog # LIEDAS217C
Supplier:   LIEDER JOHANNES
Prepared microscope slides: typical monocot roots
Catalog # LIEDAS217C
Supplier:  LIEDER JOHANNES
Prepared microscope slides: typical monocot roots
Prepared microscope slides: typical monocot roots
Convallaria (ts), lily of valley showing monocot root of endodermis, pericycle, phloem, xylem
Price
8.20 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Prepared microscope slides in systematic order showing the anatomy of typical monocot roots in angiospermae.
(ts) total structure
Specifications
  • Bezeichnung:
    Convallaria (ts), lily of valley showing monocot root of endodermis, pericycle, phloem, xylem