AuftragsverfolgungKontakt
Stoppers for Conical Ground Joints
24/29, Flat
Katalognr. 217-9095
Anbieter:   Lenz Laborglas
Stoppers for Conical Ground Joints
Katalognr. 217-9095
Anbieter:  Lenz Laborglas
Stoppers for Conical Ground Joints
Stoppers for Conical Ground Joints
24/29, Flat
Preis
9.30 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
 : DIN 12252
 : Available with flat or pointed bottom.
Technische Daten
  • NS:
    24/29
  • Typ:
    Flat
  • Seq:
    15