Stoppers for Conical Ground Joints
24/29, Flat
Katalognr. 217-9095
Anbieter: Lenz Laborglas
Direkt zu
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
- Excellent chemical resistance
- High thermal resistance, minimal heat expansion results in high resistance to temperature changes
- Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
: DIN 12252
: Available with flat or pointed bottom.
Technische Daten
- NS:24/29
- Typ:Flat
- Seq:15