Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Standard conical ground joints cones, long tip, Quickfit® | Avantor

Standard conical ground joints cones, long tip, Quickfit®

14/23 NS, 13,0 mm
Katalognr. 201-8200
Anbieter:   SciLabware
Standard conical ground joints cones, long tip, Quickfit®
Katalognr. 201-8200
Anbieter:  SciLabware
Standard conical ground joints cones, long tip, Quickfit®
Standard conical ground joints cones, long tip, Quickfit®

14/23 NS, 13,0 mm

Preis
SFr. 94.20
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents

PYREX® borosilicate glass.

  • Useful for construction of stirrer guides
  • Avoids the problem of the badge charring at high temperatures which gives an untidy appearance
  • Long tip cones manufactured with a minimum shank

Unbadged ground glass cones are free from ceramic markings.

Total length: 90 mm

Technische Daten
  • Kern:
    14/23 NS
  • Rohr A-Ø:
    13,0 mm
  • Seq:
    02