HEIZELEMENT FÜR PROLINE LAUDA 1 * 1 ST
LAUDEH201
: LAUDA
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by antigen affinity chromatography
- Antibody type:Primary
- ImmunoFluorescence:Yes
- Amino acid number:1 to 161
- Storage temperature:Cool
- Host:Rabbit
- Concentration:0,57 mg/ml
- Shipping temperature:Ice
- Antigen name:Ribosomal Protein S10
- Cat. No.:BIRBORB40873-100
- Reactivity:Human
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 161 of Human RPS10.
- Conjugation:Unconjugated
- Antigen symbol:RPS10
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).