Methyldecanoat
Glasflasche für Feststoffe, 500 mg
Katalognr. MAYA1261
Anbieter: MATREYA
Direkt zu
Product Details & Documents
Synonyme: Decanoic acid methyl ester, Capric acid methyl ester, Decylic acid methyl ester, Methyl caprate
- CAS Nummer:110-42-9
- Formel:C₁₁H₂₂O₂
- MDL-Nummer:MFCD00009580
- Molekulargewicht:186.29 g/mol
- Siedepunkt:224 °C (1013 hPa)
- Schmelzender Platingehalt:–18 °C
- Dichte:0,8724 g/cm³ (20 °C)
- Blitz Pt:94 °C
- EINECS:203-766-6
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:500 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



