Methylundecanoat
Glasflasche für Feststoffe, 100 mg
Katalognr. MAYA1166
Anbieter: MATREYA
Direkt zu
Product Details & Documents
Synonyme: Undecanoic acid methyl ester, Undecylic acid methyl ester
- CAS Nummer:1731-86-8
- Formel:C₁₂H₂₄O₂
- MDL-Nummer:MFCD00008957
- Molekulargewicht:200.32 g/mol
- EINECS:217-053-2
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:100 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



