Methyllinoleat
Glass bottle for solids, 1 g
Catalog # MAYA1025
Supplier: MATREYA
Jump to:
Product Details & Documents
Synonyms: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester
- CAS Number:112-63-0
- Formula:C₁₉H₃₄O₂
- MDL Number:MFCD00009534
- Molecular Weight:294.48 g/mol
- Boiling Pt:340 °C (1013 hPa)
- Melting Pt:–35 °C
- Density:0,89 g/cm³ (20 °C)
- EINECS:203-993-0
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



