Methyloleat
Glass bottle for solids, 1 g
Katalognr. MAYA1023
Anbieter: MATREYA
Direkt zu
Product Details & Documents
Synonyme: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
- CAS Nummer:112-62-9
- Formel:C₁₉H₃₆O₂
- MDL-Nummer:MFCD00009578
- Molekulargewicht:296.49 g/mol
- Merck-Index:12,06965
- Siedepunkt:∼218,5 °C (1013 hPa)
- Schmelzender Platingehalt:–20 °C
- Dichte:0,88 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:203-992-5
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



