Gramin
Glass bottle for solids, 500 mg
Catalog # LATOL6088-500
Supplier: LATOXAN
Jump to:
Product Details & Documents
Synonyms: 1-(1H-Indol-3-yl)-N,N-dimethylmethanamine, 3-(Dimethylaminomethyl)-Indole
- CAS Number:87-52-5
- Formula:C₁₁H₁₄N₂
- MDL Number:MFCD00005629
- Molecular Weight:174.25 g/mol
- EINECS:201-749-8
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



