Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Colchicin

Colchicin

Glass bottle for solids, 200 mg
Catalog # LATOL6051-200
Toxic
Health
Supplier:   LATOXAN
Colchicin
Catalog # LATOL6051-200
Supplier:  LATOXAN
CAS Number:  64-86-8
Colchicin

Glass bottle for solids, 200 mg

CAS Number:  64-86-8
Price
Inquire For Price
Log in for availability
1 * 200 mg
Product Details & Documents
Synonyms:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS Number:64-86-8
  • Formula:C₂₂H₂₅NO₆
  • MDL Number:MFCD00078484
  • Molecular Weight:399.44 g/mol
  • Merck Index:12,02536
  • Melting Pt:154…156 °C
  • Density:1,32 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:200-598-5
  • UN:1544
  • ADR:6.1,I
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    200 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.