AuftragsverfolgungKontakt
Biotin NTA | Avantor
Biotin NTA
Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt), 5 mg
Katalognr. 90056.
Anbieter:   Biotium
Biotin NTA
Katalognr. 90056.
Anbieter:  Biotium
CAS Nummer:  
Biotin NTA
Biotin NTA
Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt), 5 mg
CAS Nummer:  
Preis
SFr. 237.00
Einloggen für Verfügbarkeit
1 * 5 mg
Product Details & Documents
Biotinylated probes for detection of His-tagged proteins immobilised on nitrocellulose membranes.
  • White solid soluble in water
  • Removable from the histidine-tagged protein at pH 4,8
Biotin NTA, also known as BNTA (Biotin nitrilotriacetic acid, tripotassium salt) reversibly binds to histidine-tagged proteins, and can be detected with Biotium's CF™ Dye streptavidin or monoclonal anti-biotin fluorescent conjugates. Available with 6-atom aminohexanoyl (X) linker between the biotin and NTA to facilitate interaction with streptavidin.
Technische Daten
Technische Daten
  • VE:
    5 mg
  • Bezeichnung:
    Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt)
Technische Daten