Biotin NTA
Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt), 5 mg
Katalognr. 90056.
Anbieter: Biotium
Direkt zu
Product Details & Documents
Biotinylated probes for detection of His-tagged proteins immobilised on nitrocellulose membranes.
- White solid soluble in water
- Removable from the histidine-tagged protein at pH 4,8
Biotin NTA, also known as BNTA (Biotin nitrilotriacetic acid, tripotassium salt) reversibly binds to histidine-tagged proteins, and can be detected with Biotium's CF™ Dye streptavidin or monoclonal anti-biotin fluorescent conjugates. Available with 6-atom aminohexanoyl (X) linker between the biotin and NTA to facilitate interaction with streptavidin.
Technische Daten
Technische Daten
- VE:5 mg
- Bezeichnung:Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt)