Buckets, round, Azlon®
500 ml, Weiß, 17,0 L
Katalognr. 216-2810
Anbieter: SciLabware
Direkt zu
Product Details & Documents
HDPE, white or black.
- Spouted buckets for easy pouring
- Handle grip for easy lifting and carrying
- Black 10 litre bucket has metal handle for extra strength
Dokumente
AZLOBWX112_TDS_EN
Technisches Datenblatt
Technische Daten
- Kapazität:17,0 L
- Farbe:Weiß
- Teilung:500 ml