Accessories for comparators
AF332 EP colour scale, 4/64A, R1 - R7
Katalognr. TINT246410
Anbieter: Lovibond Tintometer
Direkt zu
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Technische Daten
- Messbereich:R1 - R7
- Seq:83
- Code:4/64A
- Testmethode:AF332 EP colour scale