AuftragsverfolgungKontakt
Glass beads
Glass beads
Glass beads, solid, 2 mm
Catalog # 201-0277
Glass beads
Catalog # 201-0277
Glass beads
Glass beads
Glass beads, solid, 2 mm
Price
30.60 SFr.
Log in for availability
1 * 1 kg
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifications
  • Ø:
    2 mm
  • Bezeichnung:
    Glass beads, solid
  • Seq:
    02