Glass beads
Glass beads, solid, 2 mm
Catalog # 201-0277
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifications
- Ø:2 mm
- Bezeichnung:Glass beads, solid
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.