Apotheker-Spatel
Flexible Klinge, 100 mm, 18/22 mm
RSGA051.100
: RSG
Pharmacist's spatulas with stainless steel blade and black plastic handle.
- Conical blade
- Dishwasher-proof
- Available flexible or non flexible
- Bezeichnung:Flexible Klinge
- Klingenbreite:18/22 mm
- Spitzenlänge:100 mm