Reaktionsgefäße
2,0 ml, Natur, With attached cap, Graduated, conical bottom, non sterile, RNAse free, 20000×g, 500 pieces/bag
Katalognr. BZYM710320
Marketsource
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Documents
Zertifikate
Suche nach ChargennummerTechnische Daten
- Verpackt:500 pieces/bag
- Typ:With attached cap
- Ausführung:Graduated, conical bottom, non sterile, RNAse free
- Kapazität:2,0 ml
- Farbe:Natur
- Seq:49
- RZB max.:20000×g