AuftragsverfolgungKontakt
Reaktionsgefäße
1,6 ml, With attached cap, Graduated, conical bottom, non sterile, RNAse and pyrogene free, 20000×g, 500 pieces/bag
  BZYM710318
Marketsource
 :   BIOZYM
Reaktionsgefäße
  BZYM710318
 :  BIOZYM
Marketsource
Reaktionsgefäße
1,6 ml, With attached cap, Graduated, conical bottom, non sterile, RNAse and pyrogene free, 20000×g, 500 pieces/bag
21.44 SFr.
1 * 500 ST
PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • DNase-, RNase free
With or without cap.
  • Verpackt:
    500 pieces/bag
  • Typ:
    With attached cap
  • Ausführung:
    Graduated, conical bottom, non sterile, RNAse and pyrogene free
  • Kapazität:
    1,6 ml
  • Seq:
    38
  • RZB max.:
    20000×g