Reaktionsgefäße
2,0 ml, Blau, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 500 pieces/bag
BZYM710192
Marketsource
: BIOZYM
- Verpackt:500 pieces/bag
- Typ:With attached cap
- Ausführung:Graduated, conical bottom, non sterile
- Kapazität:2,0 ml
- Farbe:Blau
- Seq:40
- RZB max.:20000×g