Reaktionsgefäße
1,6 ml, Natur, With attached cap, Graduated, conical bottom, sterile, 20000×g, 250 pieces/bag (individually wrapped )
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Dokumente
Zertifikate
Suche nach ChargennummerTechnische Daten
- Verpackt:250 pieces/bag (individually wrapped )
- Typ:With attached cap
- Ausführung:Graduated, conical bottom, sterile
- Kapazität:1,6 ml
- Farbe:Natur
- Seq:25
- RZB max.:20000×g