Reaktionsgefäße
1,6 ml, Rot, With attached cap, Graduated, conical bottom, non sterile, 20000×g, 500 pieces/bag
Katalognr. BZYM710161
Marketsource
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Documents
Zertifikate
Suche nach ChargennummerTechnische Daten
- Verpackt:500 pieces/bag
- Typ:With attached cap
- Ausführung:Graduated, conical bottom, non sterile
- Kapazität:1,6 ml
- Farbe:Rot
- Seq:31
- RZB max.:20000×g