Reaktionsgefäße
0,4 ml, Natur, With attached cap, Non graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
Katalognr. BZYM710090
Marketsource
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Zertifikate
Suche nach ChargennummerTechnische Daten
- Verpackt:1000 pieces/bag
- Typ:With attached cap
- Ausführung:Non graduated, conical bottom, non sterile
- Kapazität:0,4 ml
- Farbe:Natur
- Seq:03
- RZB max.:20000×g