Reaktionsgefäße
1,5 ml, Natur, Without cap, graduated, conical bottom, non sterile, 20000×g, 1000 pieces/bag
Katalognr. BZYM710075
Marketsource
Anbieter: BIOZYM
Direkt zu
Product Details & Documents
PE, translucent.
- Good chemical resistance
- Good temperature resistance
- DNase-, RNase free
With or without cap.
Documents
Zertifikate
Suche nach ChargennummerTechnische Daten
- Verpackt:1000 pieces/bag
- Typ:Without cap
- Ausführung:graduated, conical bottom, non sterile
- Kapazität:1,5 ml
- Farbe:Natur
- Seq:53
- RZB max.:20000×g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.