γ-(N-Morpholino)propansulfonsäure (MOPS)
Glass bottle for solids, 1 kg
Catalog # A1076.1000
Supplier: PanReac AppliChem
γ-(N-Morpholino)propansulfonsäure (MOPS)
Catalog # A1076.1000
Supplier: PanReac AppliChem
CAS Number: 1132-61-2
Jump to:
Product Details & Documents
Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- CAS Number:1132-61-2
- Formula:C₇H₁₅NO₄S
- MDL Number:MFCD00006183
- Molecular Weight:209.27 g/mol
- Merck Index:13,06288
- Melting Pt:> 250 °C
- Flash Pt:110 °C (geschlossener Tiegel)
- Storage Temperature:Raumtemperatur
- EINECS:214-478-5
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:1 kg



